Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Theobromine, 99%
Click to view available options
Quantity:
50 g
100 g
250 g
Description
Phosphodiesterase inhibitor, weak adenosine receptor antagonist, diuretic and smooth muscle relaxant.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Phosphodiesterase inhibitor, weak adenosine receptor antagonist, diuretic and smooth muscle relaxant.
Solubility
Soluble in hot methanol and dimethyl sulfoxide.
Notes
Store in cool, dry conditions in well sealed container. Store away from oxidizing agent. Store away from strong bases.
Phosphodiesterase inhibitor, weak adenosine receptor antagonist, diuretic and smooth muscle relaxant.
Solubility
Soluble in hot methanol and dimethyl sulfoxide.
Notes
Store in cool, dry conditions in well sealed container. Store away from oxidizing agent. Store away from strong bases.

Specifications
Specifications
Melting Point | 345°C to 350°C |
Density | 1.5 |
Quantity | 100 g |
Beilstein | 16464 |
Merck Index | 14,9282 |
Solubility Information | Soluble in hot methanol and dimethyl sulfoxide. |
Formula Weight | 180.17 |
Percent Purity | 99% |
Physical Form | Powder |
Chemical Name or Material | Theobromine |
Safety and Handling
Hazard Category | H302 |
Hazard Statement | GHS H Statement H301 Toxic if swallowed. |
Precautionary Statement | P264b-P270-P301+P312-P330-P501c |
EINECSNumber | 201-494-2 |
RTECSNumber | XH2275000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction