Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Theophylline Monoclonal Antibody (TH11-58.3.1.2)
GREENER_CHOICE

Catalog No. 01671958
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-671-958 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-671-958 Supplier Invitrogen™ Supplier No. MA110728
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA1-10728 detects Theophylline. MA1-10728 has been successfully used in ELISA applications. The MA1-10728 immunogen is Theophylline, conjugated to KLH.

Theophylline is a methyl xanthine derivative from tea with diuretic, smooth muscle relaxant, bronchial dilation, cardiac and central nervous system stimulant activities. Theophylline inhibits the 3',5'-cyclic nucleotide phosphodiesterase that degrades cyclic amp thus potentiates the actions of agents that act through adenylyl cyclases and cyclic amp. Theophylline is widely used in therapy of asthma and is not believed to cause liver injury. [National Center for Biotechnology Information. PubChem Compound Database; CID=2153].
TRUSTED_SUSTAINABILITY

Specifications

Antigen Theophylline
Applications ELISA
Classification Monoclonal
Clone TH11-58.3.1.2
Concentration ~1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene Alias C7H8N4O2; Dimethylxanthine; Theolair; Theophyllin
Host Species Mouse
Immunogen Theophylline, conjugated to KLH.
Purification Method purified
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Target Molecule Chemical
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.