Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Theophylline Monoclonal Antibody (TH12-104.2), QED Bioscience™

Mouse Monoclonal Antibody
Supplier: Leinco Technologies Inc 162071000UG
Description
Theophylline Monoclonal antibody specifically detects Theophylline in Chemical samples. It is validated for ELISASpecifications
| Theophylline | |
| Monoclonal | |
| 0.5-1.0 mg/mL | |
| PBS with no preservative; pH 7.4 | |
| Mouse | |
| Protein A | |
| RUO | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid | |
| Chemical |
| ELISA | |
| TH12-104.2 | |
| Unconjugated | |
| C7H8N4O2; Dimethylxanthine; Theolair; Theophyllin | |
| Theophylline-KLH. | |
| 1 mg | |
| Primary | |
| Antibody | |
| IgG1 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction