Learn More
Therapak™ 95kPa Compliant Canisters
Catalog No. 22130035 Shop All Therapak Corporation Products
Change view
Click to view available options
Quantity:
Up to 12 tubes
Up to 20 tubes
Up to 5 tubes
Size:
Small
Medium
Large
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Size
22-130-035 Up to 5 tubes Small
22-130-037 Up to 20 tubes Large
22-130-036 Up to 12 tubes Medium
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 22-130-035 Supplier Therapak™ Supplier No. 23027
Add to Cart
Add to Cart
For transport of blood tubes and urine vials and other biological substances
  • Reusable containers with durable, thick-wall construction to protect specimens
  • Ideal secondary, watertight container
  • Screw cap for secure, watertight seal
  • Small canister constructed of translucent PET to identify specimen leakage; holds up to five tubes
  • Medium canister for harsh transport conditions including dry ice shipments of frozen specimens; holds up to 6 tubes
  • Large canister is most durable of canisters and can be used for biological substances transport between client sites and reference labs; holds up to 20 tubes
Compliance
Compliant with DOT and IATA regulations as a secondary container
Specifications
Quantity Up to 5 tubes
Certifications/Compliance Comply with DOT and IATA regulations as a secondary container
Applications For transport of biological substances
Size Small
Material PET