Learn More
Therapak™ 95kPa Compliant Canisters
For transport of blood tubes and urine vials and other biological substances
$173.90 - $581.00
Specifications
Certifications/Compliance Comply with DOT and IATA regulations as a secondary container
Applications For transport of biological substances
View More Specs
Products 3
Catalog Number Mfr. No. Quantity Size Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Size Price Quantity & Availability  
22-130-037
View Documents
Therapak™
23080
Up to 20 tubes Large
Case of 6 for $394.50
Only null left
Add to Cart
22-130-035
View Documents
Therapak™
23027
Up to 5 tubes Small
Case of 48 for $581.00
Only null left
Add to Cart
22-130-036
View Documents
Therapak™
23072
Up to 12 tubes Medium
Case of 15 for $173.90
Only null left
Edge
Add to Cart
Description
Description
  • Reusable containers with durable, thick-wall construction to protect specimens
  • Ideal secondary, watertight container
  • Screw cap for secure, watertight seal
  • Small canister constructed of translucent PET to identify specimen leakage; holds up to five tubes
  • Medium canister for harsh transport conditions including dry ice shipments of frozen specimens; holds up to 6 tubes
  • Large canister is most durable of canisters and can be used for biological substances transport between client sites and reference labs; holds up to 20 tubes
Compliance
Compliant with DOT and IATA regulations as a secondary container
Specifications
Specifications
Comply with DOT and IATA regulations as a secondary container
For transport of biological substances
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.