Description
- Offer multiple temperature zones within the same container
- Specimens requiring freezing, refrigerated, and ambient temperatures can be easily segregated; required temperatures of the individual specimen can be maintained
- Interlocking foam lids help to preserve temperature; nylon mesh belt with a quick-release buckle fits across lid for added security
- Durable, easy-to-clean corrugated plastic outer shell
- Built-in reinforced metal handles assure safe transport from the vehicle to the testing area
- Available with EPS foam or high R-value urethane inserts
Order Info
Measure 28L x 19W x 14 in. D.
Specifications
Specifications
| Includes | 1/2 urethane insert |
| Applications | Large capacity courier transports |