Learn More
Therapak™ Courier Totes: Inserts
Catalog No. 22131410 Shop All Therapak Corporation Products
Change view
Click to view available options
Type:
Divider for Large EPS Insert
Divider for Large Urethane Inserts
Urethane Insert
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
22-131-410 Divider for Large Urethane Inserts
22-130-205 Urethane Insert
22-131-448 Divider for Large EPS Insert
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 22-131-410 Supplier Therapak™ Supplier No. 31021
This item is not returnable. View return policy
Replacements for use Therapak courier totes
Urethane or EPS; dividers also available.
Specifications
Certifications/Compliance Meets OSHA and DOT requirements for shipments of biological substances via ground courier transport
For Use With (Equipment) Courier Tote
Type Divider for Large Urethane Inserts