Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Therapak™ Duramark™ Fabric Lock Boxes

Catalog No. 22131427
Click to view available options
Includes:
Lock Box
Lock Box, Zipper Lock, Keys
Lock Box, Zipper Lock, Keys, Strap
Size:
Small
Large

Available in standard small and large sizes

  • Soft sides insulated with 1/4 in. foam
  • Consists of zipper closure, mesh envelope to contain refrigerant packs, and an adjustable strap for ease of courier handling during pickup activity
  • Optional zipper lock includes two matching keys, and optional security strap, which is reinforced with a steel cord sewn into the strap, helps assure the sample will be safe and secure
Standard color: Royal Blue Regency
Standard printing: Silver text with red biohazard symbol on one side only
Options include: Zipper lock, security strap
Customization: Any deviation from the standard stock configuration, which may include changes in color, size, design features (i.e. number of pockets), closure (i.e., Zipper, Velcro, buckle, etc.) and print
Minimum customization order quantity is 24 bags.
Artwork creation is available.
Small Size Dimensions: 9 x 8 x 7 in. (22.86 x 20.3 x 17.7cm)
Large Size Dimensions: 11 x 9 x 7 in. (27.9 x 22.86 x 17.7cm)

Specifications

Includes Lock Box, Zipper Lock, Keys, Strap
Color Royal Blue Regency
Size Small
Type With Red Biohazard Symbol
Material Fabric
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.