Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Therapak™ Duramark™ Fabric Lock Boxes
Available in standard small and large sizes
$143.10 - $177.10
Specifications
Color | Royal Blue Regency |
---|---|
Type | With Red Biohazard Symbol |
Material | Fabric |
Catalog Number | Mfr. No. | Includes | Size | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Includes | Size | Price | Quantity & Availability | ||||
22-131-425
|
Therapak™
36500 |
Lock Box | Small |
Each for $147.40
|
|
||||
22-131-426
|
Therapak™
36501 |
Lock Box, Zipper Lock, Keys | Small |
Each for $143.10
|
|
||||
22-131-427
|
Therapak™
36502 |
Lock Box, Zipper Lock, Keys, Strap | Small |
Each for $162.20
|
|
||||
22-131-428
|
Therapak™
36503 |
Lock Box | Large |
Each for $177.10
|
|
||||
22-131-429
|
Therapak™
36504 |
Lock Box, Zipper Lock, Keys | Large |
Each for $155.90
|
|
||||
22-131-430
|
Therapak™
36505 |
Lock Box, Zipper Lock, Keys, Strap | Large |
Each for $172.80
|
|
||||
Description
- Soft sides insulated with 1/4 in. foam
- Consists of zipper closure, mesh envelope to contain refrigerant packs, and an adjustable strap for ease of courier handling during pickup activity
- Optional zipper lock includes two matching keys, and optional security strap, which is reinforced with a steel cord sewn into the strap, helps assure the sample will be safe and secure
Standard printing: Silver text with red biohazard symbol on one side only
Options include: Zipper lock, security strap
Customization: Any deviation from the standard stock configuration, which may include changes in color, size, design features (i.e. number of pockets), closure (i.e., Zipper, Velcro, buckle, etc.) and print
Minimum customization order quantity is 24 bags.
Artwork creation is available.
Small Size Dimensions: 9 x 8 x 7 in. (22.86 x 20.3 x 17.7cm)
Large Size Dimensions: 11 x 9 x 7 in. (27.9 x 22.86 x 17.7cm)
Specifications
Royal Blue Regency | |
With Red Biohazard Symbol | |
Fabric |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title