Learn More
Therapak™ IATA Shipping Boxes for Ambient Transport
Ideal kit packing boxes for reference labs, clinical trials and pathology kits
Supplier:  Therapak™ 23409G
 View more versions of this product
Catalog No. 22-131-488
This item is not returnable. View return policy
Description
Description
  • Each box has a specific tab-lock mechanism to keep boxes closed during transport
  • Ambient shipping boxes ideal for kit packaging and outbound shipping of diagnostic kits
  • Known as two-way shipper boxes, all ambient shipper boxes can be used for return transport to the laboratory in addition to outbound shipping
Compliance
Meets IATA Packing Instruction 650 dimensional requirements.
Specifications
Specifications
Meets IATA Packing Instruction 650 dimensional requirements.
Large
11.25 x 10.75 x 4.25 in.
For shipment of Biological Substance Category B specimens
Side-Load IATA Shipping Box
11 x 10.5 x 4 in.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.