Learn More
Description
- 24 Gauge cold rolled steel construction
- Painted with powder coat finish for excellent strength and durability
- Insulated on inside with foam assisting preserving the integrity of the specimens
- With hinged front door that rotates down 90° for convenient access and use as temporary tray
- Dimensions: 14 × 10 1/2 × 7 in.
- Supplied with lock and two matching keys for security
- Standard color: White
- Standard printing: Reflex blue text, red biohazard symbol on front side only
- Optional straps: Under the door, over the door
- Customization: Any deviation from the standard stock configuration, which may include changes in color, size, design features, closure and print
Specifications
Specifications
| Includes | Metal Lock Box + Under Door Strap |
| Material | 24 gauge cold rolled steel construction |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.