Learn More
Therapak™ Plastic Duramark™ Lock Boxes
Catalog No. 22131450 Shop All Therapak Corporation Products
Change view
Click to view available options
Includes:
Lock Box
Lock Box with Over Door Strap
Lock Box with Under Door Strap
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
22-131-450 Lock Box with Under Door Strap
22-130-206 Lock Box with Over Door Strap
22-131-452 Lock Box
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 22-131-450 Supplier Therapak™ Supplier No. 36743
Add to Cart
Add to Cart
High impact polystyrene construction
  • Made for hanging on the door using the optional straps
  • Provides the timesaving convenience of a viewing window and a hinged door that opens down for ease of use
  • Comes with lock and two matching keys for adding security
  • Dimensions: 13.5L x 10.25W x 6.75 in. H
  • Standard color: Off-white
  • Standard printing: Reflex blue text, red biohazard symbol on front side only
  • Optional straps: Under the door, over the door
  • Customization: Any deviation from the standard stock configuration, which may include changes in color, size, design features, closure and print
Specifications
Includes Lock Box with Under Door Strap
Color Off-White
Material High-impact Polystyrene