Learn More
Therapak™ Single Specimen Collection Kits
Catalog No. 22131552 Shop All Therapak Corporation Products
Change view
Click to view available options
Includes:
Evidence Seal
Temp. Strip, Evidence Seal, Bag
Temperature Strip
Temperature Strip, Evidence Seal
Temperature Strip, Transport Bag
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
22-131-552 Evidence Seal
22-131-551 Temperature Strip
22-131-553 Temperature Strip, Evidence Seal
22-131-555 Temperature Strip, Transport Bag
22-131-554 Temp. Strip, Evidence Seal, Bag
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 22-131-552 Supplier Therapak™ Supplier No. 40003
Add to Cart
Add to Cart
This item is not returnable. View return policy
Single Specimen Collection Kits can be used in conjunction with on-site testing products or remote site collection when specimen is transported to a laboratory for analysis.
Single Specimen Collection Kits can be used in conjunction with on-site testing products or remote site collection when specimen is transported to a laboratory for analysis. Each kit contains either a 180 mL beaker style collection cup with pour spout or a leak-resistant 90 mL sterile container with cap and integrity seal. The transport bag features two sealable, tamper evident pockets and comes with absorbent material. The individually bagged kits are available in various configurations.
Specifications
Product Type Urine Specimen Container
Sterility Sterile
For Use With (Application) For drugs of abuse testing, on-site testing or remote site collection
Includes Evidence Seal
Volume (English) 3 oz.
Volume (Metric) 90 mL