Globe Scientific Thermal Printer Paper for Clinical Chemistry Analyzers

Thermal printer paper for use with popular chemistry analyzers.

Manufacturer: Globe Scientific 7155

 View more versions of this product

Catalog No. 21-200-108

  Check Availability
Add to Cart

    Specifications

    For Use With (Equipment) Roche Cobas Mira analyzer, Tosoh 360 analyzers
    Packaging Quantity 10 rolls/box; 48 rolls/case
    Description 80mm wide x 47mm dia. x 82 ft. long
    Product Type Printer Paper

    • For use with Roche™ Cobas Mira™ analyzer, Tosoh™ 360 analyzers
    • Size (L x W): 82 ft. x 80mm
    • Dia.: 47mm
    • Quantity: 10 rolls/box; 48 rolls/case

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit