Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Interscience Thermal Transfer Film



Click to view available options
For Use With (Equipment):
DataLink Pro
Thermal Printer
Description
- Thermal transfer printer ribbon
- Available for Interscience thermal printers or DataLink Pro
Specifications
Specifications
Product Type | Thermal Transfer Printer Ribbon |
Color | Black |
For Use With (Equipment) | Thermal Printer |
Print Technology | Thermal Transfer |
Autoclavable | Not Autoclavable |
Sterility | Non-sterile |
Format | Roll |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction