Learn More
Thermo Scientific™ 48-Well Semi-Skirted Plates
Catalog No. ab0648y
Click to view available options
Color:
Yellow
48-well semi-skirted plate for PCR and qPCR applications.
The Thermo Scientific 48-well semi-skirted plate for PCR and qPCR applications comes in a convenient format, which allows for lower sample numbers. The semi skirt makes the plate more rigid and allots space for labeling and barcoding. Available in assorted colors, it's compatible with standard multichannel pipettes and automated liquid handling systems.The plate is sealable when used with adhesive films, foils or cap strips.
Highlights
• Convenient format for lower sample numbers
• Semi-skirt adds rigidity and allows for labeling or barcoding
• Sealable using adhesive films and foils and flat, ultra clear and domed cap strips
• 0.3 mL maximum well volume (when used with adhesive and heat seals)
TRUSTED_SUSTAINABILITY
Specifications
Format Standard
Array 6 x 8
Color Yellow
Material Virgin Polypropylene
No. of Wells 48
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.