Learn More
Thermo Scientific™ Adapters for Thermo Scientific™ 24 x 1.5/2.0 mL Rotor with ClickSeal Biocontainment Lid
Description
Increase your flexibility for micro-volume separations using adapters for our standard microcentrifuge rotor. Select from a number of Microliter Tube Adapters and PCR adapters for the Thermo Scientific™ 24 x 1.5/2.0mL Rotor with ClickSeal Biocontainment Lid.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter.
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Specifications
Type | Microtube Adapter |
Tube Type | Microtube |
Capacity | 24 x 0.5/0.6 mL Microliter Tubes |
For Use With (Equipment) | 24 x 1.5/2.0mL Rotor with ClickSeal Biocontainment Lid |
Max. RCF | 21100 |
Max. Speed | 14800 |
Tube Size | 0.6/0.5 mL |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.