Learn More
Thermo Scientific™ BetaBasic™ 18 DASH-HTS HPLC Columns, 5μm Particle Size
Designed for high throughput, these short columns are excellent for ballistic gradients, with retention and sensitivity for very fast LC/MS applications
Manufacturer:  Thermo Scientific™ 71505022150
 View more versions of this product
Catalog No. 71505022150
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • Patented, low dead volume design minimizes band spreading
  • Finger-tight fittings make changing columns quick and easy
Features of BetaBasic 18 Solid Phase
  • Highly efficient and reproducible columns
  • Pore size suitable for small molecules, peptides and protein digests
  • Superb pH stability
Specifications
Specifications
DASH-HTS Column, 1/Pk.
20 mm
2 to 12
200 m²/g
C18
13%
Yes
2.1 mm
5 μm
150 Å
L1
Silica
Reversed Phase
Reversed Phase