Learn More
Thermo Scientific™ BetaBasic™ CN CPF HPLC Columns
Description
Thermo Scientific HPLC columns are manufactured under strict ISO 9001:2000 procedures, and every batch of silica and bonded phase is thoroughly tested for physical and chromatographic characteristics. Test reports are included with every column shipped.
- For small molecules, peptides and protein digests
- Low pH stability compared to other cyano phases
- Offer alternative selectivity based on dipole-dipole and dipole-induced dipole interactions, and can be used in normal or reversed phase separations
Specifications
Specifications
| Diameter (Metric) | 3 mm |
| Length (Metric) | 250 mm |
| Particle Size | 5 μm |
| pH | 2 to 9 |
| Pore Size | 150 Å |
| Surface Area | 200 m²/g |
| USP Type | L10 |
| Stationary Phase | Cyano (CN) |
| Packing Material | Silica |
| Carbon Load | 5% |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.