Learn More
Fisherbrand™ Castaloy™ Adjustable-Angle Clamps
Two- or three-prong clamps are good for gripping rods up to 0.75 in. (19mm) diameter
 View more versions of this product
Catalog No. 05-769-5Q
Includes:
Vinyl and fiberglass sleeves
Description
Description
Two- or three-prong clamps are good for gripping rods up to 0.75 in. (19mm) diameter
  • Two-prong flat jaws are best for test tubes while three-prong are suited for holding irregular shapes
  • Holding angle of jaw is adjustable, locked with a wingnut
  • Wingnut also allows length adjustment
Specifications
Specifications
1 Ea.
0.5 in.
5.7 cm
2-Prong
Angle Clamp
1.27 cm
2.25 in.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.