Learn More
Thermo Scientific™ Clear Strong Heat Sealing Film
Description
- Clear polyester polypropylene laminate heat seal
- Strong and unpeelable
- Also suitable for storage and disposal of hazardous material
- Good solvent resistance; recommended for use with DMSO
- Seal integrity range: -80°C to +120°C
Specifications
Specifications
| Color | Clear |
| Sterility | Non-sterile |