Learn More
Epredia™ Permafrost™ Slides
Click to view available options
Color:
Aqua
Blue
Gold
Green
Lavender
Orange
Pink
Red
Tan
White
Yellow
Description
- Resistant to commonly used laboratory solvents
- 1.0 to 1.2 mm thick with ground edges
Specifications
Specifications
Frosted | Permafrost 1 End |
Material | Glass |
Thickness (Metric) | 1 to 1.2 mm |
Edges | Ground |
Color | Pink |
Quantity | 1440/Cs. |