Learn More
Thermo Scientific™ DNA Quantitation Standards
Catalog No. p-2855941
Click to view available options
:
40 micrograms
40micrograms; supplied in ReddyRun Buffer
500bp; 40micrograms; supplied in ReddyRun Buffer
Includes
Standards supplied in 50μg/mL ReddyRun™ Buffer include a separate vial of 10X ReddyRun Buffer to add to samples.
Ideal for incorporation into samples as internal quantitation controls
  • Offer a choice of fragment sizes
  • Concentration of each standard: 100μg/mL
  • Provide versatile quantitation for ethidium bromide stained gels
  • Mirror the effect seen in samples—larger fragments tend to form sharper bands and smaller fragments more diffuse bands in agarose gels
  • Supplied in TE storage buffer
Fragment sizes:
QSK-101, QSK-101L: 200 bp, 500 bp, and 1000 bp plus two prelabeled tubes
QS1000, QS1000LD: 1000bp
QS200, QS200LD: 2000bp
QS500LD: 500bp
TRUSTED_SUSTAINABILITY
Specifications
Product Type Dilution and Delivery Device System
Quantity 250/Cs.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.