Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Lysis Buffer for MagJET RNA Kit

Catalog No. ferr2302
Click to view available options
Quantity:
200 mL

Catalog No. FERR2302

Supplier: Thermo Scientific™ R2302

Generate sample lysate in order to isolate total RNA. Thermo Scientific™ MagJET™ Lysate Buffer is a component of the MagJET™ RNA Kits (K2731/2732) and may be purchased separately when additional solution is needed. It is provided in 200mL bottles.

The Thermo Scientific™ MagJET™ RNA Kit is designed for fast and efficient purification of total RNA from cultured mammalian cells, tissues, bacteria, and yeast. The kit utilizes paramagnetic bead technology, enabling high yields and robust performance. High binding capacity, uniform particle size, and rapid magnetic response of MagJET magnetic beads makes the technology ideal for high throughput automatic nucleic acid purification, as well as for manual purification by low sample throughput users. The resulting high-quality RNA is free of proteins, nucleases and other contaminants or inhibitors.

TRUSTED_SUSTAINABILITY

Specifications

Buffer Lysis Buffer
Product Line MagJET
Quantity 200 mL
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.