Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Luminaris HiGreen qPCR Master Mix, low ROX

Catalog No. p-4861499
Change view
Click to view available options
No. of Reactions:
1250 Reactions
250 Reactions
500 Reactions
5000 Reactions
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
FERK0972 500 Reactions
FERK0971 250 Reactions
FERK0974 5000 Reactions
FERK0973 1250 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. FERK0972 Supplier Thermo Scientific™ Supplier No. K0972
Only null left
Add to Cart
Add to Cart

Includes

Luminaris HiGreen Low ROX qPCR Master Mix (2X) containing Hot Start Taq DNA Polymerase, UDG, dNTPs (with dUTP) and SYBR Green I in an optimized PCR buffer. The master mix contains low concentration of ROX passive reference dye for specific real-ti

Thermo Scientific Luminaris Color HiGreen and Luminaris HiGreen qPCR Master Mixes are universal ready-to-use solutions optimized for qPCR and two-step RT-qPCR.

  • UDG in the master mix to prevent carry-over contamination
  • Equal performance of blue and colorless master mixes
  • Versions with premixed ROX or fluorescein for different real-time platforms
  • Standard cycling protocols
  • Excellent qPCR performance
  • Specificity: Hot Start Taq DNA Polymerase and the optimized buffer eliminate non-specific amplification and formation of primer dimers
  • Sensitivity: single copy detection
  • Wide linear range: accurate quantification across 9 orders of magnitude
  • Reproducibility and convenience: ready-to-use 2X master mix
TRUSTED_SUSTAINABILITY

Specifications

Concentration 2X
Content And Storage Contains:
• 4 x 1.25 mL Luminaris HiGreen Low ROX qPCR Master Mix (2X) containing Hot Start Taq DNA Polymerase, UDG, dNTPs (with dUTP) and SYBR Green I in an optimized PCR buffer; sufficient for 500 x 20 μL reactions. The master mix contains low concentration of ROX passive reference dye for specific real-time platforms.
• 4 x 1.25 mL Nuclease-Free Water

Store at -20°C.
Detection Method SYBR
GC-Rich PCR Performance High
PCR Method qPCR
Polymerase Taq DNA Polymerase
Reaction Speed Standard
For Use With (Equipment) 7500 System, QuantStudio™, ViiA™ 7 System, Stratagene Mx4000, Stratagene Mx3000P, Stratagene Mx3005P
Product Line Luminaris
Product Type qPCR Master Mix
Quantity 500 reactions
Sufficient For 500 Reactions
For Use With (Application) Gene Expression
Fidelity (vs. Taq) 2 X
No. of Reactions 500 Reactions
Sample Type DNA, dsDNA
Volume 7.5 mL
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.