Learn More
Thermo Scientific™ MagJET™ Plasmid DNA Kit
Purifies plasmid DNA from overnight E. coli cultures using paramagnetic bead technology.
Manufacturer:  Thermo Scientific™ K2791
 View more versions of this product
Catalog No. FERK2791
Add to cart
Description
Description
The Thermo Scientific MagJET Plasmid DNA Kit is designed for fast and efficient purification of plasmid DNA (up to 4-5 µg/mL) or high copy plasmid DNA from overnight E. coli culture.
the kit utilizes paramagnetic bead technology enabling high yields and robust performance. High binding capacity, uniform particle size, and rapid magnetic response of MagJET Magnetic Beads makes the technology ideal for high throughput automatic nucleic acid purification, as well as for manual purification for low sample throughput users.
The resulting high quality DNA is free of proteins, nucleases and other contaminants or inhibitors. Purified plasmid DNA can be used in a wide range of downstream applications such as PCR, qPCR and other enzymatic reactions.
Highlights
·High yields - up to 5 µg of plasmid DNA from 1 mL of bacterial culture
·Pure - isolated DNA is immediately ready-to-use in downstream application such as sequencing, transcription, digestion with restriction enzymes or transfection of robust cell lines
·Flexible - automation and manual processing protocols available
·Repeatable results and robust performance
Applications
Fast isolation of high purity plasmid DNA suitable for all conventional molecular biology procedures, including:
· FastDigest or conventional restriction digestion
· Automated fluorescent and radioactive sequencing
· PCR
· In vitro transcription
· Transformation
· Transfection of robust cell lines
Includes
· Lysis Buffer
· Resuspension Buffer
· Neutralization Buffer
· MagJET Magnetic Beads
· RNase A
· Wash Buffer 1 (conc.)
· Wash Buffer 2 (conc.)
· Elution Buffer
Hazardous:Yes
> Hazardous::No
> Quality Control:Thermo Scientific MagJET Plasmid DNA Kit is qualified by isolating high copy plasmid DNA from 0.5-5 mL of overnight E. coli culture grown in LB or rich growth medium (OD600 = 2-6). The quality of isolated DNA is evaluated spectrophotometrically and by agarose gel electrophoresis. The purified DNA has an A260/A280 ratio of 1.8±0.2.
> Shelf Life:36 Months
> Shipping Condition:Ambient
> Storage Condition:Ambient
Specifications
Specifications
High purity plasmid DNA suitable for all conventional molecular biology procedures like: FastDigest or conventional restriction digestion, Automated fluorescent and radioactive sequencing, PCR, In vitro transcription, Transformation, Transfection of robust cell lines
Lysis Buffer, Resuspension Buffer, Neutralization Buffer, MagJET Magnetic Beads, RNase A, Wash Buffer 1 (conc.), Wash Buffer 2 (conc.), Elution Buffer
For 96 Preparations
Once opened store RNase A solution at -20deg.C. MagJET Magnetic Beads should be stored at 4deg.C. Other components of the kit should be stored at room temperature (15deg. To 25deg.C).
Safety and Handling
Safety and Handling
ShelfLife : 3 Years