Learn More
Thermo Scientific™ Reagents for MagJET™ gDNA Kit
Catalog No. p-4861475
Click to view available options
Quantity:
22 mL
25 mL
90 mL
The Thermo Scientific™ MagJET™ kits are designed for fast and efficient purification of nucleic acid. The kits utilize paramagnetic bead technology, enabling high yields and robust performance. High binding capacity, uniform particle size, and rapid magnetic response of MagJET magnetic beads makes the technology ideal for high throughput automatic nucleic acid purification, as well as for manual purification by low sample throughput users. The resulting high-quality DNA is free of proteins, nucleases and other contaminants or inhibitors.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) MagJET gDNA Kit
Quantity 22 mL
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.