Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ GeneJET™ Plasmid Maxiprep Kit Reagents

Catalog No. FERR2271
Click to view available options
Product Type:
Elution Buffer
Endotoxin Binding Reagent
Lysis Solution
Neutralization Solution
Resuspension Solution
Wash Buffer I (conc.)
Wash Buffer II (conc.)

Includes

Wash Solution II (concentrated)

Reagents for use with the GeneJET Plasmid Maxiprep Kit. The Plasmid Maxiprep Kit is designed for large scale isolation of high quality plasmid DNA from recombinant E. coli cultures.

GeneJET Plasmid Maxiprep Kit utilizes silica-based membrane technology in the form of convenient spin column. Each prep recovers up to 750μg of high-copy plasmid DNA that can be used in a wide variety of molecular biology procedures, such as restriction endonuclease digestion, PCR, cloning, transformation, automated sequencing, in vitro transcription, and transfection of robust cell lines.

The Thermo Scientific GeneJET Plasmid Maxiprep Kit utilizes silica-based membrane technology in the form of convenient spin column. Each prep recovers up to 750 μg of high copy plasmid DNA that can be used in a wide variety of molecular biology procedures, such as restriction endonuclease digestion, PCR, cloning, transformation, automated sequencing, in vitro transcription, and transfection of robust cell lines.

Highlights

  • Efficient - high yields up to 750 μg of high quality plasmid DNA from 250 mL of bacterial culture
  • High concentration of DNA purified - > 400 ng/μL
    Convenient - spin column format, centrifuge or vacuum protocols
  • Fast - procedure takes 60 minutes, considerably faster than using gravity flow columns
Applications
  • Isolated plasmid DNA is suitable for all conventional molecular biology procedures, including:
  • Transfection of robust cell lines
  • FastDigest restriction enzyme digestion
  • Automated fluorescent sequencing
  • PCR
  • Cloning
  • Transformation
  • In vitro transcription

Hazardous: No

Quality Control:

  • The GeneJET Plasmid Maxiprep Kit is qualified by isolating high copy number plasmid DNA from 250 mL of overnight E. coli culture (OD600 = 2) grown in LB medium following the protocol outlined in the manual.
  • The kit passes QC requirements if the purified plasmid DNA has an A260/280 ratio between 1.8 and 1.9 and a dominant band of supercoiled plasmid DNA is observed after agarose gel electrophoresis.
  • The functional quality of the plasmid DNA is evaluated by digestion with restriction endonucleases.
Storage Condition: Ambient

 

Isolated plasmid DNA is suitable for all conventional molecular biology procedures, including:

  • Transfection of robust cell lines
  • FastDigest restriction enzyme digestion
  • Automated fluorescent sequencing
  • PCR
  • Cloning
  • Transformation
  • In vitro transcription

 

Order Info

Orders placed Monday through Wednesday ship next day; Thursday and Friday orders ship the following Monday.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at room temperature
For Use With (Equipment) GeneJET™ Endo-free Plasmid Maxiprep Kit
Product Type Wash Buffer II (conc.)
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.