Learn More
Thermo Scientific™ GLP Compliance Services for Plasma Blast Freezer
Catalog No. p-3704628
Change view
Click to view available options
Product Type:
GLP Compliance Services for Plasma Blast Freezer
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
13-262-249 GLP Compliance Services for Plasma Blast Freezer
13-262-212 -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 13-262-249 Supplier Thermo Scientific™ Supplier No. IOQPCKE89003549
Only null left
Add to Cart
Add to Cart
Request a Quote
Stay compliant with our industry exculsive no charge requalificaiton guarantee included with each Thermo Scientific XBF
IQ
  • Verifies that the equipment, manuals, supplies and any other accessories arrived undamaged as specified in the sales order. Verifies equipment is assembled and installed and any initial diagnostics tests have been performed. Any abnormal event(s) observed during assembly and installation have been documented.
  • Utility, Facility and Environment Verifies that the installation site satisfactorily meets manufacturer-specified environmental requirements.
OQ System Component Information Detailed configuration information for each system component have been recorded.
  • OQ Limits: Record a list of manufacturer recommended limits for ensuring that the system is operating as expected.
  • Equipment Operational Tests and Results: Test important equipment functions to verify that the equipment operates as intended by the manufacturer and required by the user. This is a group of selected important equipment parameters for testing according to the intended use of the equipment.
Key Tests: Door Ajar, Power Failure, Temperature Alerts
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Thermo Scientific Blast Freezer including Thermo Scientific XBF
Description Validation (IQ/OQ) includes a standardized protocol procedure, IQ: Verify Installation and environmental conditions, OQ: Verify operation to design specification, perform empty chamber temperature mapping, open door and power loss testing.
Product Type GLP Compliance Services for Plasma Blast Freezer
Location Contact your local sales representative for a list of standard and extended coverage areas.
Includes Includes travel to major metropolitan areas. Does not include Installation. Instrument must be installed powered up and set temperature point for 24 hours before on-site service