Learn More
Thermo Scientific™ Hypersil BDS CN CPG HPLC Columns--3μm Particle Size
Supplier:  Thermo Scientific™ 28803103031
 View more versions of this product
Catalog No. 28803103031
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Specifications
Specifications
3 mm
3 μm
130 Å
L10
Silica
Mixed Mode
100 mm
2 to 9
170 m²/g
Cyano (CN)
4%
Normal and Reversed Phase