Learn More
Thermo Scientific™ Hypersil™ Duet C18/SAX Mixed-Mode Ion Exchange CPG Columns
Catalog No. 34305151031
Change view
Click to view available options
Particle Size: 5μm; 100L x 1.0mm I.D.
Particle Size: 5μm; 100L x 2.1mm I.D.
Particle Size: 5μm; 100L x 3.0mm I.D.
Particle Size: 5μm; 100L x 4.0mm I.D.
Particle Size: 5μm; 100L x 4.6mm I.D.
Particle Size: 5μm; 125L x 2.1mm I.D.
Particle Size: 5μm; 125L x 3.0mm I.D.
Particle Size: 5μm; 125L x 4.0mm I.D.
Particle Size: 5μm; 125L x 4.6mm I.D.
Particle Size: 5μm; 150L x 1.0mm I.D.
Particle Size: 5μm; 150L x 2.1mm I.D.
Particle Size: 5μm; 150L x 3.0mm I.D.
Particle Size: 5μm; 150L x 4.0mm I.D.
Particle Size: 5μm; 150L x 4.6mm I.D.
Particle Size: 5μm; 250L x 1.0mm I.D.
Particle Size: 5μm; 250L x 2.1mm I.D.
Particle Size: 5μm; 250L x 3.0mm I.D.
Particle Size: 5μm; 250L x 4.0mm I.D.
Particle Size: 5μm; 250L x 4.6mm I.D.
Particle Size: 5μm; 30L x 2.1mm I.D.
Particle Size: 5μm; 30L x 3.0mm I.D.
Particle Size: 5μm; 30L x 4.6mm I.D.
Particle Size: 5μm; 50L x 1.0mm I.D.
Particle Size: 5μm; 50L x 2.1mm I.D.
Particle Size: 5μm; 50L x 3.0mm I.D
Particle Size: 5μm; 50L x 4.6mm I.D.
26 product options available for selection
Product selection table with 26 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
34305151031 Particle Size: 5μm; 150L x 1.0mm I.D.
34305032131 Particle Size: 5μm; 30L x 2.1mm I.D.
34305033031 Particle Size: 5μm; 30L x 3.0mm I.D.
34305034631 Particle Size: 5μm; 30L x 4.6mm I.D.
34305051031 Particle Size: 5μm; 50L x 1.0mm I.D.
34305052131 Particle Size: 5μm; 50L x 2.1mm I.D.
34305053031 Particle Size: 5μm; 50L x 3.0mm I.D
34305054631 Particle Size: 5μm; 50L x 4.6mm I.D.
34305101031 Particle Size: 5μm; 100L x 1.0mm I.D.
34305102131 Particle Size: 5μm; 100L x 2.1mm I.D.
34305103031 Particle Size: 5μm; 100L x 3.0mm I.D.
34305104031 Particle Size: 5μm; 100L x 4.0mm I.D.
34305104631 Particle Size: 5μm; 100L x 4.6mm I.D.
34305122131 Particle Size: 5μm; 125L x 2.1mm I.D.
34305123031 Particle Size: 5μm; 125L x 3.0mm I.D.
34305124031 Particle Size: 5μm; 125L x 4.0mm I.D.
34305124631 Particle Size: 5μm; 125L x 4.6mm I.D.
34305152131 Particle Size: 5μm; 150L x 2.1mm I.D.
34305153031 Particle Size: 5μm; 150L x 3.0mm I.D.
34305154031 Particle Size: 5μm; 150L x 4.0mm I.D.
34305154631 Particle Size: 5μm; 150L x 4.6mm I.D.
34305251031 Particle Size: 5μm; 250L x 1.0mm I.D.
34305252131 Particle Size: 5μm; 250L x 2.1mm I.D.
34305253031 Particle Size: 5μm; 250L x 3.0mm I.D.
34305254031 Particle Size: 5μm; 250L x 4.0mm I.D.
34305254631 Particle Size: 5μm; 250L x 4.6mm I.D.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 26 options available.
26 options
Catalog No. 34305151031 Supplier Thermo Scientific™ Supplier No. 34305151031