Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Klenow Fragment of DNA Polymerase I

Catalog No. AB0268
Change view
Click to view available options
Quantity:
1000 Units
5000 Units
Concentration:
5 U/μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Concentration
AB-0268 1000 Units 5 U/μL
AB-0268B 5000 Units 5 U/μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AB-0268 Supplier Thermo Scientific™ Supplier No. AB0268

Includes

Separate 100mM MgCl2

Removes overhanging 3' ends; fills in DNA fragments with overhanging 5'ends

  • Storage and Dilution Buffer: 50mM KH2PO2 (pH 7.0 at 25°C), 1mM DTT, 50% (v/v) glycerol
  • Reaction Buffer: 5X concentration. 250mM KH2PO2 (pH 7.0 at 25°C), 5mM DTT

TRUSTED_SUSTAINABILITY

Specifications

Concentration 5 U/μL
Quantity 1000 Units
Product Type DNA Polymerase I
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.