Learn More
Thermo Scientific™ MAS™ LabLink xL COMMUNICATOR and CLOUD COMMUNICATOR
Description
Automatically upload QC data from various clinical laboratory instruments, middlewares and laboratory information systems to the Thermo Scientific™ MAS™ LabLink xL WEB.
Automatically upload QC data from various clinical laboratory instruments, middlewares and laboratory information systems to the Thermo Scientific™ MAS™ LabLink xL WEB with Thermo Scientific™ MAS™ LabLink xL COMMUNICATOR and CLOUD COMMUNICATOR. Real-time, fully-automated and semi-automated connectivity options are available, all of which will streamline QC data transfer protocol, reduce costs and eliminate transcription errors.
- Fully automated, real time QC data transfer via Data Innovations Instrument Manager™ and Beckman REMISOL Advance™ data manager to LabLink xL
- Numerous semi-automated connectivity options available for QC data upload from instrument, LIS or middleware to LabLink xL
- Simple to setup
Specifications
Specifications
| Description | LabLink xL COMMUNICATOR and CLOUD COMMUNICATOR |
| Sample Type | Various |
| Technology | Data Transfer |
| Type | Communicator |