Learn More
Thermo Scientific™ Microplate Readers Optical Filters
Description
- All filters have a 3nm bandpass centered at the specified wavelength
- Compatible with all three Multiskan models
Specifications
Specifications
| Wavelength | 595 nm |