Learn More
Thermo Scientific™ Nunc™Flasks, Non-treated
SCI_ED
Catalog No. S04852
Click to view available options
:
Non-treated EasYFlask 175cm2, Filter Cap, total volume 645mL, Working Volume 55mL, 30/Cs
This item is not returnable. View return policy
This item is not returnable. View return policy
Includes
Flask with cap
For cells that do not require Nunclon™ Δ treated surface.
Nunc non-treated cell culture flasks are made of high quality optically clear polystyrene for suspension cell culture.
  • White caps for easy identification
  • Angled neck for easy access
  • EasyFlasks with ergonomic filter cap allows constant airflow and minimizes chance of contamination
  • TripleFlask with three parallel layers and footprint of a standard 175cm2 flask same external dimensions
  • Sterilized by gamma irradiation and certified to be non-pyrogenic
Order Info
Extra caps in every case.
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 55 mL
Material Polystyrene
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.