Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Orion™ Basic Electrodes
SCI_ED

Catalog No. s04954
Click to view available options
Connector Types:
BNC
This item is not returnable. View return policy
This item is not returnable. View return policy

For basic pH measurements of common samples

Low maintenance gel-filled reference system for simplified use and storage

  • 0-14pH range
  • 0-80°C temperature range for pH electrodes
  • Can be used with any meter with BNC input

TRUSTED_SUSTAINABILITY

Specifications

Connector Types BNC
Material Epoxy
pH Range 0 to 14
Temperature (Metric) Operating 0°C to 80°C
For Use With (Equipment) Russell meters
Storage Requirements Store in Orion pH electrode storage solution 910001
Temperature (English) Operating 32°F to 176°F
Temperature (Metric) Range 0°C to 80°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.