Learn More
Thermo Scientific™ Owl™ ADJ1 Adjustable System Accessories and Replacement Parts
Description
- The height of the upper buffer chamber (UBC) may be adjusted by sliding the UBC up or down two support poles allowing gels to vary in height from 14 cm to 40 cm.
- The notched or offset glass plate of the gel sandwich presses against a silicone gasket in the UBC for a leakproof seal.
- Plates are clamped to the UBC with either binder clamps or heavy duty pony clamps.
Specifications
Specifications
| For Use With (Equipment) | Owl ADJ1 Adjustable Electrophoresis System |
| Type | Notched Glass Plate |
| Material | Glass |
| Thickness (English) | 0.18 in. |
| Thickness (Metric) | 0.48 cm |
| Dimensions (L x W) | 9.8 x 6.2 in. (25 x 16 cm) |
| Length (English) | 9.8 in. |
| Length (Metric) | 25 cm |
| Width (English) | 6.2 in. |
| Width (English) Gel | 6.2 in. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.