Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Owl™ ADJ1 Adjustable System Accessories and Replacement Parts

Catalog No. OWADJ1C40
Change view
Click to view available options
Type:
Blank Glass Plate
Gasket
Notched Glass Plate
Offset Glass Plate
Spacer Set
Dimensions (L x W):
14.9 x 6.2 in. (38 x 16 cm)
15.7 x 0.39 in. (40 x 1 cm)
15.7 x 6.2 in. (40 x 16 cm)
9.05 x 6.2 in. (23 x 16 cm)
9.8 x 0.39 in. (25 x 1 cm)
9.8 x 6.2 in. (25 x 16 cm)
Thickness (English):
0.03 in.
0.05 in.
0.18 in.
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type Dimensions (L x W) Thickness (English)
OW-ADJ1C40 Spacer Set 15.7 x 0.39 in. (40 x 1 cm) 0.03 in.
09-528-169K Blank Glass Plate 15.7 x 6.2 in. (40 x 16 cm) 0.18 in.
OW-ADJ123G Offset Glass Plate 9.05 x 6.2 in. (23 x 16 cm) 0.18 in.
09-528-169L Offset Glass Plate 14.9 x 6.2 in. (38 x 16 cm) 0.18 in.
OW-ADJ140R Notched Glass Plate 15.7 x 6.2 in. (40 x 16 cm) 0.18 in.
09-528-170F Spacer Set 9.8 x 0.39 in. (25 x 1 cm) 0.03 in.
OW-ADJ1GK Gasket - -
09-528-170E Spacer Set 9.8 x 0.39 in. (25 x 1 cm) 0.05 in.
OW-ADJ1D40 Spacer Set 15.7 x 0.39 in. (40 x 1 cm) 0.05 in.
09-528-169F Notched Glass Plate 9.8 x 6.2 in. (25 x 16 cm) 0.18 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Catalog No. OW-ADJ1C40 Supplier Thermo Scientific™ Supplier No. ADJ1C40

Extremely compact at only 16cm wide

  • The height of the upper buffer chamber (UBC) may be adjusted by sliding the UBC up or down two support poles allowing gels to vary in height from 14 cm to 40 cm.
  • The notched or offset glass plate of the gel sandwich presses against a silicone gasket in the UBC for a leakproof seal.
  • Plates are clamped to the UBC with either binder clamps or heavy duty pony clamps.

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) Owl ADJ1 Adjustable Electrophoresis System
Type Spacer Set
Includes One bottom spacer and two side spacers
Thickness (English) 0.03 in.
Thickness (Metric) 0.08 cm
Dimensions (L x W) 15.7 x 0.39 in. (40 x 1 cm)
Length (English) 15.7 in.
Length (Metric) 40 cm
Width (English) 0.39 in.
Width (English) Gel 6.3 in.
Width (Metric) 1 cm
Width (Metric) Gel 16 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.