Learn More
Thermo Scientific™ Owl™ Adjustable Vertical Electrophoresis Systems
Catalog No. 09528168
Change view
Click to view available options
Gel size: 16 x 14-40cm; Buffer volume: 600mL; L x W: 23 x 25cm
Gel size: 35 x 20-80cm; Buffer volume: 800mL; L x W: 32 x 35cm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
09-528-168 Gel size: 16 x 14-40cm; Buffer volume: 600mL; L x W: 23 x 25cm
OW-ADJ3 Gel size: 35 x 20-80cm; Buffer volume: 800mL; L x W: 32 x 35cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 09-528-168 Supplier Thermo Scientific™ Supplier No. ADJ1
Includes
Removable UBC with safety interlocking lid and attached power supply lead, Removable LBC with safety interlocking lid with attached power supply lead; Base with attached pole rings; Two heavy-duty poles
  • The height of the upper buffer chamber (UBC) may be adjusted by sliding the UBC up or down two support poles
  • Thumbscrews on the ring assembly secure the UBC in the desired position
  • Ring assemblies on the base of the apparatus secure the poles upright
  • The notched or offset glass plate of the gel sandwich presses against a silicone gasket on the UBC for a leakproof seal
  • Glass plates are clamped to the UBC with binder clamps or heavy duty pony clamps
  • The unit can be dismantled for easy cleaning and storage