Learn More
Thermo Scientific™ Owl OSP Electrophoresis Power Supplies
Description
- Built-in timer permits unattended runs
- Four outputs allow four units to be run with clear LED readout
- Safety features include no-load detection, current/load alarm, and sudden load detection
- Samples are protected by automatic restart after power failure
- 300V unit ideal for DNA, RNA, mini SDS-page, pulse field electrophoresis, tank electroblotting, and electroelution
- 500V unit suited for DNA, RNA, and protein electrophoresis
Specifications
Specifications
| Model | OSP-300 Lightning Volt |
| Voltage | 10-300 V, in 1 V steps |