Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Oxoid™ IMAGEN RSV

Catalog No. rk6102112
Click to view available options
Quantity:
50 Tests/Kit
This item is not returnable. View return policy
This item is not returnable. View return policy

For the direct detection of respiratory syncytial virus (RSV) in clinical specimens

  • Test utilizes three monoclonal antibodies to detect specific structural proteins expressed in all strains of human RSV

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2°C to 8°C
Sample Type Clinical samples such as nasopharyngeal aspirates
Includes 2 x 1 well Positive Control Slide; 3 mL of Mounting Fluid; 1.4 mL of IMAGEN RSV Reagent
Quantity 50 Tests/Kit
Analysis Time 30 min.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.