Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Pierce™ Anti-HA Agarose
GSA_VA

Catalog No. p-4332113
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI26181 1 mL
PI26182 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PI26181 Supplier Thermo Scientific™ Supplier No. 26181
Only null left
Add to Cart
Edge
Add to Cart

HA Epitope Tag Monoclonal antibody 26181 and 26182 from Thermo Scientific™ specifically detects HA Epitope Tag in Tag samples, and it is validated for Immunoprecipitation

An HA tag is frequently incorporated into recombinant proteins for a variety of purposes. An anti-HA antibody can then be used to detect the protein when doing studies with transfected cells.
Host Species: Mouse
Cross Reactivity: Tag
Isotype: IgG1
Clone: 2-2.2.14
Immunogen: HA peptide YPYDVPDYA derivitized to ovalbumin
Formulation: 1mL Purified Antibody in PBS, with 0.05% sodium azide; pH7.2
Synonyms: human influenza hemagglutinin, HA-tag, HA, HA Epitope Tag
26181 is supplied as 1mL of antibody on crosslinked 4% beaded agarose supplied as a 50% slurry in 0.1M phosphate, 0.15M NaCl, pH 7.2 with 0.05% sodium azide. Specific for the HA peptide YPYDVPDYA. The anti-HA agarose has been used successfully in immunoprecipitation applications. The anti-HA agarose has 3.5mg mouse anti-HA IgG1 monoclonal antibody conjugated permL of settled agarose resin. HA Synthetic Peptide (Product # 26184) is available for use in neutralization and control experiments and to competitively elute HA-tagged fusion proteins from immobilized anti-HA affinity resin.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Product Type Agarose
Column Type Agarose Resin, Affinity
Format Bottle
Conjugate HA-epitope tag (YPYDVPDYA)
Purification Target Anti-HA Antibody
Content And Storage 1 mL of settle resin, provided as 2 mL of 50% slurry. Store at 2–8°C.
Quantity 1 mL
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.