Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ B-PER™ Complete Bacterial Protein Extraction Reagent
GSA_VA

Catalog No. p-133431
Click to view available options
Quantity:
250 mL
500 mL

Catalog No. PI89821

Supplier: Thermo Scientific™ 89821

Only null left
Add to Cart
Edge
Add to Cart

Includes

Reagent packages are available in Tris and phosphate buffer formulations; Kits contain Tris-based B-PER Reagent plus lysozyme and DNase I enzymes.

Efficient, gentle lysis and extraction of E.coli and other bacterial cells

  • An easy-to-use cell lysis reagent that effectively disrupts cells and solubilizes native or recombinant proteins without denaturation
  • All B-PER Reagents are compatible with downstream applications such as affinity chromatography (e.g., immobilized metal affinity chromatography, glutathione chromatography), SDS-PAGE, and protein quantification (e.g., Pierce BCA Protein Assay, Pierce 660nm Protein Assay)
  • This all-inclusive formulation combines the lysis reagent with lysozyme, and a universal nuclease to enable mild extraction from both gram-negative and gram-positive bacteria
  • Depending on the particular application protease inhibitors, salts, reducing agents, denaturants, and chelating agents may be added to the reagent.

Features of the B-PER Bacterial Protein Extraction Reagent:

  • Detergent in Tris buffer with Lysozyme and Universal Nuclease
  • Improved cell lysis and DNA digestion, thereby releasing soluble proteins and reducing viscosity to increase yields
  • Recovery of large molecular weight proteins (high MW proteins)
  • Cost-effective equivalent to Merck Millipore Bugbuster™ Master Mix
  • More effective than traditional sonication and typical homemade lysis buffers, many of which include detergents and components that interfere with downstream applications

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Formulation Proprietary detergent in 20mM Tris buffer, pH 7.5, containing lysozyme and universal nuclease
Quantity 250 mL
Product Type Bacterial Protein Extraction Reagent
Volume (Metric) 250 mL
Content And Storage Store at 4°C.
Product Line B-PER
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.