Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Pierce™ c-Myc-Tag Magnetic IP/Co-IP Kit

Catalog No. pi88844
Click to view available options
Quantity:
40 reactions kit

Magnetic beads for efficient isolation of c-Myc-tagged proteins and interactors.

The Thermo Scientific Pierce Magnetic c-Myc-Tag IP/Co-IP Kit contains specific immunoaffinity magnetic beads and reagents to perform immunoprecipitation assays of c-Myc fusion proteins or co-IP experiments using c-Myc-tagged bait proteins.

Unlike traditional IP procedures based on capture with Protein A/G beads, this kit uses magnetic beads containing pre-immobilized anti-c-Myc antibody. These Pierce Anti-c-Myc Magnetic Beads ensure specific binding of c-Myc tagged protein complexes from biological samples. Because the antibody is covalently attached to the beads, IP-targets are easily eluted and recovered without antibody contamination. The complete IP kit includes the magnetic beads, lysis/wash buffer, low-pH elution buffer, neutralization buffer, c-Myc-tag positive control lysate, and non-reducing sample buffer for SDS-PAGE. Protocols are provided for both manual and automated magnetic separation workflows. Sufficient components are provided to perform 40 IP or co-IP assays.

Features of the c-Myc-Tag Magnetic IP/Co-IP Kit:

  • Specific magnetic beads—covalently immobilized high-quality anti-c-Myc monoclonal antibody enables high yields of immunoprecipitation products
  • Low non-specific binding—stable, pre-blocked beads and specific antibody minimize off-target binding
  • Trouble-free elution—low-pH elution buffer ensures recovery of c-Myc-tagged protein interaction complexes without antibody leaching contamination
  • Convenient and fast—complete kit and easy-to-follow instructions provide optimized protocols to perform IP or Co-IP experiment in approximately 1 hour
  • Versatile—magnetic beads are compatible with manual and automated magnetic separation workflows (e.g., Thermo Scientific KingFisher Instruments)

The c-Myc peptide (EQKLISEEDL) derived from the C-terminus region of human c-Myc protein is one of several fusion protein tags used for recombinant protein expression. The Pierce c-Myc Magnetic IP/Co-IP Kit uses a specific, high-affinity immobilized antibody (clone 9E10) for rapid immunoprecipitation of c-Myc tagged fusion proteins from bacterial and mammalian cell lysates, as well as from lysates prepared with the Pierce Human in vitro Translation Kits. The beads are incubated with a cell lysate containing c-Myc tagged protein, the fusion protein is captured, and the beads are subsequently washed and then eluted using low-pH elution buffer or non-reducing sample buffer. The protocol and buffers have been optimized for both IP and co-IP reactions, enriching for specific protein interaction complexes in the eluted samples. Anti-c-Myc antibody can be used to detect c-Myc tagged protein by Western blot analysis.

For information about the binding capacity and other properties of the magnetic beads used in this kit, see Pierce Anti-c-Myc Magnetic Beads (Part No. 88842).

Related Products

Pierce™ c-Myc-Tag IP/Co-IP Kit (Part No. 23620)

TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 1 mL
Formulation Slurry: 10 mg/mL, 1% solids
Content And Storage Sufficient For: 40 IP reactions using 25 μL of anti-c-Myc magnetic beads
• c-Myc-tagged Positive Control (Part No. 23633), 500 μL
• Pierce Anti-c-Myc Magnetic Beads, 1 mL
• Mag c-Myc IP/Co-IP Buffer-1, 50 mL
• Mag c-Myc IP/Co-IP Buffer-2 (20X), 20 mL
• Non-Reducing Sample Buffer (5X), 5 mL
• Elution Buffer (pH 2.0), 5 mL

Store at 4°C.
Assay Affinity Purification
For Use With (Equipment) KingFisher™ Flex Magnetic Particle Processor, Magnetic Stand
Product Line Pierce™
Quantity 40 reactions kit
For Use With (Application) Pull-Down (Tag-Based), Immunoprecipitation (IP)
Target Target-Specific
Type Tag IP/Co-IP Kit
Sufficient For 40 Reactions
Format Kit
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.