Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Cathepsin D, Recombinant Protein, Invitrogen™

Catalog No. PIRP38944
Click to view available options
Quantity:
10 μg

Recombinant Protein

Cathepsin D Recombinant Protein, RP-38944, from Invitrogen.

Cathepsin D is synthesized as a 54kDa precursor, which is proteolytically processed to an intermediate 48kDa single chain, which matures into more stable 34kDa and 14kDa two chain form. It is an astrogen-regulated lysosomal protease that has been suggested to facilitate cancer cell migration and invasion by diagesting the basement membrane, extracellular matrix, and connective tissue. Because of its mitogenic and proteolytic activities, it has been implicated as a prognostic marker in many tumor types. Cathespin D is expressed in epitthelial cells as well as in macrophages. This product is a recombinant protein corresponding to full-length human cathepsin D. On SDS-PAGE commassie blue stained gel, the purified recombinant protein shows a band at 48kDa.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P07339
Concentration 0.1mg/mL
For Use With (Application) Control
Formulation 0.05M tris HCl with 0.01M L-glutathione and no preservative; pH 7.5
Gene ID (Entrez) 1509
Name Human Cathepsin D
pH Range 7.5
Quantity 10 μg
Source E. coli
Storage Requirements -20°C or -80°C if preferred
Regulatory Status RUO
Common Name Human Cathepsin D
Gene Symbol CTSD
Specific Reactivity Human
Conjugate Unconjugated
Cross Reactivity Hu
Species E. coli
Recombinant Recombinant
Form Liquid
Protein Subtype Recombinant
Purity or Quality Grade purified
Show More Show Less

in vitro research only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.