Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Pierce™ D-Luciferin, Monopotassium Salt

Catalog No. p-135223
Click to view available options
Quantity:
100 mg
1 g

Firefly luciferase substrate for in vitro and in vivo applications

Thermo Scientific Pierce D-Luciferin is formulated at greater than 99% purity as both monosodium and monopotassium salts to provide convenience and high performance at a low cost in reporter assays involving firefly luciferase.

Luciferin is the substrate for bioluminescence reactions with firefly luciferase enzyme. Lyophilized potassium and sodium salts of firefly luciferin are stable and easily soluble, and both formulations are compatible with a wide variety of luciferase-based in vivo imaging and in vitro bioluminescent assays. (The decision to use one or the other form is a matter of researcher preference.) Applications include ATP assays, gene reporter assays, high-throughput sequencing and various contamination assays throughout the biotechnology field. For in vivo imaging, the injection of luciferin in model systems enables real-time, non-invasive monitoring of disease progression and/or drug efficacy through bioluminescence.

Highlights:

  • High purity, typically greater than 99.7%, as verified by HPLC and FTIR
  • Great performance with in vitro applications
  • Suitable for in vivo imaging
Applications:
  • Gene reporter assays
  • ATP assays
  • High throughput sequencing
  • Contamination assays
  • Bioluminescent in vivo imaging
  • Drug screening
  • Multiplexed assays to study off-target effects
  • Signal transduction pathway analysis

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at or below -20°C.
Detection Method Bioluminescence
For Use With (Application) ATP Assay, Gene Reporter Assay
Product Type D-Luciferin, Monopotassium Salt
Product Line Pierce™
Quantity 100 mg
Substrate Properties Chemical Substrate
Substrate Type Luciferase Substrate

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.