Learn More
Thermo Scientific™ Pierce™ DyLight 547 and 647 Monoclonal Ab Labeling Kits
Catalog No. PI53015
Click to view available options
Quantity:
1 Kit
Specifically developed for fast, efficient labeling of 100ug of monoclonal antibody and exceptional antibody recovery
The DyLight Dyes are an excellent alternative to Cy3 and Cy5 Dyes, providing greater photostability and fluorescence over a broad range of pH values. DyLight 547 replaces Cy3; DyLight 647replaces Cy5.
  • Label and purify antibodies in approximately one hour Amine reactive dyes label virtually any antibody Fluorescent dye pre-measured eliminates the time and waste associated with weighing dye Samples containing as low as 20ug/mL can be processed with unsurpassed recovery and ≥95% retention of small molecules (<1,000 Da) Average antibody recovery of 80% Highly efficient free dye removal Zeba Spin Column format eliminates the need for column preparation, screening fractions or waiting for antibody to elute from column
  • DyLight Dye 547
    Excitation 557nm Emission 570nm Extinction Coefficient (min) 150,000 M-1 cm-1
    DyLight Dye 647
    Excitation 652nm Emission 673nm Extinction Coefficient (min) 250,000 M-1 cm-1
TRUSTED_SUSTAINABILITY
Specifications
Quantity 1 Kit
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.