Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human IFN gamma, Recombinant Protein, Invitrogen™

Catalog No. PIRP75631
Click to view available options
Quantity:
1 mg

Recombinant Protein

Interferon gamma (human) Recombinant Protein, RP-75631, from Invitrogen.

RP-75631 contains purified recombinant human IFN gamma protein with an N-terminal hexahistidine tag. RP-75631 may be used as a positive control in many immunological assays.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P01579
For Use With (Application) Control, Western Blot
Formulation PBS with 50% glycerol and no preservative
Gene ID (Entrez) 3458
Name Human IFN gamma
Quantity 1 mg
Source E. coli
Immunogen N/A
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Common Name Human IFN gamma
Gene Symbol IFNG
Specific Reactivity Human
Conjugate Unconjugated
Cross Reactivity Hu
Species E. coli
Recombinant Recombinant
Form Liquid
Protein Subtype Recombinant
Purity or Quality Grade purified
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.