Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Involucrin, Recombinant Protein, Invitrogen™

Catalog No. PIRP39142
Click to view available options
Quantity:
10 μg

Recombinant Protein

Involucrin Recombinant Protein, RP-39142, from Invitrogen.

Involucrin is a soluble cytoplasmic protein precursor of the epidermal cornfied envelope that becomes cross-linked by transglutaminase during envelope assembly. Involucrin is expressed in a range of stratified squamous epithelia, including the cornea which lacks a distinct cornfied layer. The protein is a useful marker of terminal differentiation. It is tightly linked to the onset of differentiation and first expressed in the immediate suprabasal layers of the epidermis. This product is a GST-tagged recombinant protein corresponding to human involucrin aa 420-578. On SDS-PAGE commassie blue stained gel, the purified recombinant protein shows a band at 41kDa (including GST).
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P07476
Concentration 0.1mg/mL
For Use With (Application) Control
Formulation 0.05M tris HCl with 0.01M L-glutathione and no preservative; pH 7.5
Gene ID (Entrez) 3713
Name Human Involucrin
pH Range 7.5
Quantity 10 μg
Source E. coli
Immunogen N/A
Storage Requirements -20°C or -80°C if preferred
Regulatory Status RUO
Common Name Human Involucrin
Gene Symbol IVL
Specific Reactivity Human
Conjugate Unconjugated
Cross Reactivity Hu
Species E. coli
Recombinant Recombinant
Form Liquid
Protein Subtype Recombinant
Purity or Quality Grade purified
Show More Show Less

in vitro research only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.