Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ Pierce™ MMP-18, N-Terminal
Applications: ELISA, IHC, IP, WB
Supplier: Thermo Scientific™ OPA108180
Description
Host Species: Rabbit
Cross Reactivity: Hu
Immunogen: Synthetic peptide based on the N-terminal region of human MMP-18.
ELISA, Immunohistochemistry, Immunoprecipitation, Western Blotting

Specifications
MMP-18 | |
1mg/mL | |
Rabbit | |
Affinity Purified | |
Primary |
Polyclonal | |
Unconjugated | |
Synthetic peptide based on the N-terminal region of human MMP-18. | |
100 μg | |
Human |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction